missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-nitrobenzoic Acid, TCI America™

Manufacturer:  TCI America F039925G

 View more versions of this product

Catalog No. F039925G



Specifications

Specifications

4-Fluoro-3-nitrobenzoic Acid
121°C
453-71-4
MFCD00007058
BOJWTAQWPVBIPG-UHFFFAOYSA-N
185.11
185.11
≥98.0% (GC,T)
Yellow
25g
C7H4FNO4
4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])F
67987
Crystal-Powder at 20°C

Chemical Identifiers

453-71-4
185.11
BOJWTAQWPVBIPG-UHFFFAOYSA-N
67987
C7H4FNO4
MFCD00007058
4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])F
Safety and Handling

Safety and Handling

TSCA : No

Documents