Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-nitrobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-nitrobenzeneboronic acid pinacol ester
CAS 939968-60-2
Formula Weight 267.06
MDL Number MFCD12546515
Beilstein 20289173
Synonym 4-Fluoro-3-nitrophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6272003

 
alfa aesar™
H6272003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6272006

 
alfa aesar™
H6272006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62720MD

 
alfa aesar™
H62720MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.06
Melting Point 64° to 65°C
Chemical Name or Material 4-Fluoro-3-nitrobenzeneboronic acid pinacol ester
Physical Form Crystalline Solid
Assay 96%
MDL Number MFCD12546515
CAS 939968-60-2
Beilstein 20289173
Synonym 4-Fluoro-3-nitrophenylboronic acid pinacol ester
Storage Note 1 2° to 8°C
Color Yellow
Quantity 1g

Applications
4-Fluoro-3-nitrobenzeneboronic acid is generally used as a intermediate in pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Safety and Handling

2° to 8°C