Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-nitrobenzaldehyde, 98%

$39.32 - $145.70

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-nitrobenzaldehyde
CAS 42564-51-2
Molecular Formula C7H4FO3
Formula Weight 169.11
MDL Number MFCD01861388
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2618103

  SDS
alfa aesar™
H2618103
1g Each for $39.32

AAH2618106

  SDS
alfa aesar™
H2618106
5g Each for $145.70
Description & Specifications

Specifications

Formula Weight 169.11
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
CAS 42564-51-2
Flash Point >110°C (230°F)
Quantity 1g
Health Hazard 1 Warning
Chemical Name or Material 4-Fluoro-3-nitrobenzaldehyde
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H4FO3
MDL Number MFCD01861388
Melting Point 44° to 47°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.