missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-nitroaniline, TCI America™

Manufacturer:  TCI America F016825G

 View more versions of this product

Catalog No. F016825G



Specifications

Specifications

4-Fluoro-3-nitroaniline
97°C
364-76-1
MFCD00007833
LLIOADBCFIXIEU-UHFFFAOYSA-N
4-fluoro-3-nitroaniline
67768
156.12
≥98.0% (GC)
Undesignated
25g
C6H5FN2O2
3-nitro-4-fluoroaniline, 4-fluoro-3-nitrobenzenamine, benzenamine, 4-fluoro-3-nitro, aniline, 4-fluoro-3-nitro, 4-fluoro-3-nitro-aniline, 5-amino-2-fluoronitrobenzene, 4-fluoro-3-nitro-phenylamine, 3-nitro-4-fluoroanailine, 3-nitro-4-fluoro aniline, 4-fluoro-3-nitro aniline
C1=CC(=C(C=C1N)[N+](=O)[O-])F
156.116
CHEBI:48642
Crystal-Powder at 20°C

Chemical Identifiers

364-76-1
156.116
LLIOADBCFIXIEU-UHFFFAOYSA-N
67768
4-fluoro-3-nitroaniline
C6H5FN2O2
MFCD00007833
3-nitro-4-fluoroaniline, 4-fluoro-3-nitrobenzenamine, benzenamine, 4-fluoro-3-nitro, aniline, 4-fluoro-3-nitro, 4-fluoro-3-nitro-aniline, 5-amino-2-fluoronitrobenzene, 4-fluoro-3-nitro-phenylamine, 3-nitro-4-fluoroanailine, 3-nitro-4-fluoro aniline, 4-fluoro-3-nitro aniline
CHEBI:48642
C1=CC(=C(C=C1N)[N+](=O)[O-])F
Safety and Handling

Safety and Handling

RTECSNumber : BY1700000

TSCA : Yes

Documents