missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-3-nitroaniline 98.0+%, TCI America™
$56.41 - $56.41
Chemical Identifiers
CAS | 364-76-1 |
---|---|
Molecular Formula | C6H5FN2O2 |
Molecular Weight (g/mol) | 156.12 |
MDL Number | MFCD00007833 |
InChI Key | LLIOADBCFIXIEU-UHFFFAOYSA-N |
Synonym | 3-nitro-4-fluoroaniline, 4-fluoro-3-nitrobenzenamine, benzenamine, 4-fluoro-3-nitro, aniline, 4-fluoro-3-nitro, 4-fluoro-3-nitro-aniline, 5-amino-2-fluoronitrobenzene, 4-fluoro-3-nitro-phenylamine, 3-nitro-4-fluoroanailine, 3-nitro-4-fluoro aniline, 4-fluoro-3-nitro aniline |
PubChem CID | 67768 |
ChEBI | CHEBI:48642 |
IUPAC Name | 4-fluoro-3-nitroaniline |
SMILES | NC1=CC=C(F)C(=C1)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
F016825G
|
TCI America
F016825G |
25 g |
Each for $56.41
|
|
|||||
Chemical Identifiers
364-76-1 | |
156.12 | |
LLIOADBCFIXIEU-UHFFFAOYSA-N | |
67768 | |
4-fluoro-3-nitroaniline |
C6H5FN2O2 | |
MFCD00007833 | |
3-nitro-4-fluoroaniline, 4-fluoro-3-nitrobenzenamine, benzenamine, 4-fluoro-3-nitro, aniline, 4-fluoro-3-nitro, 4-fluoro-3-nitro-aniline, 5-amino-2-fluoronitrobenzene, 4-fluoro-3-nitro-phenylamine, 3-nitro-4-fluoroanailine, 3-nitro-4-fluoro aniline, 4-fluoro-3-nitro aniline | |
CHEBI:48642 | |
NC1=CC=C(F)C(=C1)[N+]([O-])=O |
Specifications
364-76-1 | |
C6H5FN2O2 | |
25 g | |
LLIOADBCFIXIEU-UHFFFAOYSA-N | |
4-fluoro-3-nitroaniline | |
67768 | |
156.12 | |
Crystalline Powder |
97°C | |
MFCD00007833 | |
3-nitro-4-fluoroaniline, 4-fluoro-3-nitrobenzenamine, benzenamine, 4-fluoro-3-nitro, aniline, 4-fluoro-3-nitro, 4-fluoro-3-nitro-aniline, 5-amino-2-fluoronitrobenzene, 4-fluoro-3-nitro-phenylamine, 3-nitro-4-fluoroanailine, 3-nitro-4-fluoro aniline, 4-fluoro-3-nitro aniline | |
NC1=CC=C(F)C(=C1)[N+]([O-])=O | |
156.12 | |
CHEBI:48642 | |
≥98.0% (GC) | |
4-Fluoro-3-nitroaniline |
Safety and Handling
RTECSNumber : BY1700000
TSCA : Yes