Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoro-3'-nitroacetophenone, 98%

$38.12 - $102.15

Chemical Identifiers

Chemical Name or Material 4'-Fluoro-3'-nitroacetophenone
CAS 400-93-1
Molecular Formula C8H6FNO3
Formula Weight 183.14
MDL Number MFCD00115369
Beilstein 2211976
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1989903

  SDS
alfa aesar™
L1989903
1g Each for $38.12

AAL1989906

  SDS
alfa aesar™
L1989906
5g Each for $102.15
Description & Specifications

Specifications

Formula Weight 183.14
Chemical Name or Material 4'-Fluoro-3'-nitroacetophenone
Assay 98%
Molecular Formula C8H6FNO3
MDL Number MFCD00115369
CAS 400-93-1
Beilstein 2211976
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Boiling Point 118° to 120°C (0.8mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Melting Point 45° to 50°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.