missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoro-3'-nitroacetophenone, 98%

Manufacturer:  Alfa Aesar™ L1989903

 View more versions of this product

Catalog No. AAL1989903



Specifications

Specifications

4'-Fluoro-3'-nitroacetophenone
Odorless
45°C to 50°C
400-93-1
MFCD00115369
1-4-fluoro-3-nitro-phenyl ethanone
CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-]
183.138
183.14
>110°C (230°F)
118°C to 120°C (0.8mmHg)
1g
C8H6FNO3
2211976
PTCNZDJJIOLIKQ-UHFFFAOYSA-N
1-(4-fluoro-3-nitrophenyl)ethanone
2734612
98%

Chemical Identifiers

400-93-1
183.138
PTCNZDJJIOLIKQ-UHFFFAOYSA-N
2734612
CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-]
C8H6FNO3
MFCD00115369
1-4-fluoro-3-nitro-phenyl ethanone
1-(4-fluoro-3-nitrophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents