missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-methylphenylboronic acid, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC435330250

 View more versions of this product

Catalog No. AC435330250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Fluoro-3-methylphenylboronic acid
97.5
White
97.5% min. (HPLC)
C7H8BFO2
Glass bottle
25g
B(C1=CC(=C(C=C1)F)C)(O)O
153.95
153.95
98%
139911-27-6
100.0
215.4°C to 217.3°C
Authentic
MFCD01863527
3-methyl-4-fluorophenylboronic acid, 4-fluoro-3-methylbenzeneboronic acid, 4-fluoro-3-methylphenyl boronic acid, 4-fluoro-3-methylphenylboronicacid, 4-fluoro-m-tolylboronic acid, 5-borono-2-fluorotoluene, acmc-209cjo, boronic acid, 4-fluoro-3-methylphenyl, pubchem5146
JCIJCHSRVPSOML-UHFFFAOYSA-N
(4-fluoro-3-methylphenyl)boronic acid
2774580
Crystalline Powder

Chemical Identifiers

139911-27-6
153.95
JCIJCHSRVPSOML-UHFFFAOYSA-N
2774580
B(C1=CC(=C(C=C1)F)C)(O)O
C7H8BFO2
MFCD01863527
3-methyl-4-fluorophenylboronic acid, 4-fluoro-3-methylbenzeneboronic acid, 4-fluoro-3-methylphenyl boronic acid, 4-fluoro-3-methylphenylboronicacid, 4-fluoro-m-tolylboronic acid, 5-borono-2-fluorotoluene, acmc-209cjo, boronic acid, 4-fluoro-3-methylphenyl, pubchem5146
(4-fluoro-3-methylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only