missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-methylphenol, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC311950050

 View more versions of this product

Catalog No. AC311950050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Fluoro-3-methylphenol
5g
Authentic
1.5125 to 1.5145 (20°C, 589nm)
≥97.5%
CH3C6H3(F)OH
MFCD00042296
Clear colorless to pale yellow
452-70-0
Glass bottle
Liquid
C7H7FO
126.13
4-Fluoro-m-cresol

Chemical Identifiers

C7H7FO
126.13
4-Fluoro-m-cresol
≥97.5%
CH3C6H3(F)OH
MFCD00042296
4-Fluoro-3-methylphenol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.