missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-3-methylphenol, Thermo Scientific™
Chemical Identifiers
CAS | 452-70-0 |
---|---|
Molecular Formula | C7H7FO |
Molecular Weight (g/mol) | 126.13 |
MDL Number | MFCD00042296 |
InChI Key | RVYGYYVGWSCWGY-UHFFFAOYSA-N |
Synonym | 2-fluoro-5-hydroxytoluene, 3-methyl-4-fluoro phenol, 3-methyl-4-fluorophenol, 4-fluoro-3-methyl phenol, 4-fluoro-3-methyl-phenol, 4-fluoro-3-methylphenol, 4-fluoro-m-cresol, acmc-209k1i, phenol, 4-fluoro-3-methyl, pubchem1501 |
PubChem CID | 2737394 |
IUPAC Name | 4-fluoro-3-methylphenol |
SMILES | CC1=C(C=CC(=C1)O)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC311950010
|
Thermo Fisher Scientific
AC311950010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
452-70-0 | |
126.13 | |
RVYGYYVGWSCWGY-UHFFFAOYSA-N | |
2737394 | |
CC1=C(C=CC(=C1)O)F |
C7H7FO | |
MFCD00042296 | |
2-fluoro-5-hydroxytoluene, 3-methyl-4-fluoro phenol, 3-methyl-4-fluorophenol, 4-fluoro-3-methyl phenol, 4-fluoro-3-methyl-phenol, 4-fluoro-3-methylphenol, 4-fluoro-m-cresol, acmc-209k1i, phenol, 4-fluoro-3-methyl, pubchem1501 | |
4-fluoro-3-methylphenol |
Specifications
452-70-0 | |
32.0°C | |
Authentic | |
CH3C6H3(F)OH | |
Glass bottle | |
2-fluoro-5-hydroxytoluene, 3-methyl-4-fluoro phenol, 3-methyl-4-fluorophenol, 4-fluoro-3-methyl phenol, 4-fluoro-3-methyl-phenol, 4-fluoro-3-methylphenol, 4-fluoro-m-cresol, acmc-209k1i, phenol, 4-fluoro-3-methyl, pubchem1501 | |
RVYGYYVGWSCWGY-UHFFFAOYSA-N | |
4-fluoro-3-methylphenol | |
2737394 | |
Liquid | |
4-Fluoro-3-methylphenol |
Colorless to Yellow | |
97°C | |
C7H7FO | |
MFCD00042296 | |
1.5125 to 1.5145 (20°C, 589nm) | |
1g | |
CC1=C(C=CC(=C1)O)F | |
126.13 | |
126.13 | |
≥97.5% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
RUO – Research Use Only