Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methylphenol, 98%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-methylphenol
CAS 452-70-0
Molecular Formula C7H7FO
Formula Weight 126.13
MDL Number MFCD00042296
UN Number UN3261
Synonym 4-Fluoro-m-cresol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1989703

 
alfa aesar™
L1989703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1989706

 
alfa aesar™
L1989706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1989714

 
alfa aesar™
L1989714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.13
Chemical Name or Material 4-Fluoro-3-methylphenol
Assay 98%
CAS 452-70-0
Synonym 4-Fluoro-m-cresol
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Molecular Formula C7H7FO
MDL Number MFCD00042296
UN Number UN3261
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Flash Point 96°C (204°F)
Odor Phenol-like
Boiling Point 76°C (5mmHg)
Melting Point 28° to 32°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.