Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methylbenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-methylbenzyl bromide
CAS 261951-70-6
Molecular Formula C8H8BrF
Formula Weight 203.06
MDL Number MFCD01631425
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2668103

 
alfa aesar™
H2668103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2668106

 
alfa aesar™
H2668106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.06
Chemical Name or Material 4-Fluoro-3-methylbenzyl bromide
Molecular Formula C8H8BrF
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 80°C (3mmHg)
Quantity 1g
Assay 97%
MDL Number MFCD01631425
CAS 261951-70-6

Applications
Pharmaceutical Intermediate. Organic synthesis, tear gas mixture of ortho-, meta-, and para-isomers.

Solubility
Insoluble in water.

Notes
Lachrymatory. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases. Store protected from moisture.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.