Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methylbenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-methylbenzeneboronic acid pinacol ester
CAS 1001200-60-7
Formula Weight 236.09
MDL Number MFCD18729904
Synonym 4-Fluoro-3-methylphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6271703

 
alfa aesar™
H6271703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62717MD

 
alfa aesar™
H62717MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.09
Assay 96%
Synonym 4-Fluoro-3-methylphenylboronic acid pinacol ester
Chemical Name or Material 4-Fluoro-3-methylbenzeneboronic acid pinacol ester
Physical Form Solid
Quantity 1g
CAS 1001200-60-7
MDL Number MFCD18729904