Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methylbenzeneboronic acid, 98%

$89.51 - $89.51

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-methylbenzeneboronic acid
CAS 139911-27-6
Molecular Formula C7H8BFO2
Formula Weight 153.95
MDL Number MFCD01863527
Beilstein 7474834
Synonym 4-Fluoro-3-methylphenylboronic acid; 4-Fluoro-m-tolylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1875306

  SDS
alfa aesar™
L1875306
5g Each for $89.51

AAL1875314

  SDS
alfa aesar™
L1875314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.95
Chemical Name or Material 4-Fluoro-3-methylbenzeneboronic acid
Assay 98%
MDL Number MFCD01863527
Health Hazard 1 Warning
CAS 139911-27-6
Beilstein 7474834
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 208° to 211°C
Quantity 5g
Molecular Formula C7H8BFO2
Synonym 4-Fluoro-3-methylphenylboronic acid; 4-Fluoro-m-tolylboronic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.