Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methylbenzaldehyde, 97%

$52.03 - $164.70

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-methylbenzaldehyde
CAS 135427-08-6
Molecular Formula C8H7FO
Formula Weight 138.14
MDL Number MFCD01631431
Beilstein 4797794
Synonym 4-Fluoro-m-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2352603

  SDS
alfa aesar™
B2352603
1g Each for $52.03

AAB2352606

  SDS
alfa aesar™
B2352606
5g Each for $164.70
Description & Specifications

Specifications

Formula Weight 138.14
Chemical Name or Material 4-Fluoro-3-methylbenzaldehyde
Assay 97%
Molecular Formula C8H7FO
MDL Number MFCD01631431
CAS 135427-08-6
Beilstein 4797794
Synonym 4-Fluoro-m-tolualdehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 208°C
Quantity 1g

Applications
4-Fluoro-3-methylbenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff field.

Solubility
Slightly soluble in water.

Notes
Store in refrigerator at 4°C. Incompatible material is strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.