Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methylanisole, 98%

$45.41 - $153.93

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-methylanisole
CAS 2338-54-7
Molecular Formula C8H9FO
Formula Weight 140.16
MDL Number MFCD00042293
Beilstein 2080015
Synonym 2-Fluoro-5-methoxytoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1869406

 
alfa aesar™
A1869406
5g Each for $45.41

AAA1869414

 
alfa aesar™
A1869414
25g Each for $153.93
Description & Specifications

Specifications

Formula Weight 140.16
Chemical Name or Material 4-Fluoro-3-methylanisole
Molecular Formula C8H9FO
CAS 2338-54-7
Beilstein 2080015
Synonym 2-Fluoro-5-methoxytoluene
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 62°C (143°F)
Boiling Point 174° to 175°C
Assay 98%
MDL Number MFCD00042293
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.