Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoro-3'-methylacetophenone, 97%

$57.09 - $229.35

Chemical Identifiers

Chemical Name or Material 4'-Fluoro-3'-methylacetophenone
CAS 369-32-4
Molecular Formula C9H9FO
Formula Weight 152.17
MDL Number MFCD04972879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3249903

 
alfa aesar™
H3249903
1g Each for $57.09

AAH3249906

 
alfa aesar™
H3249906
5g Each for $229.35
Description & Specifications

Specifications

Formula Weight 152.17
Chemical Name or Material 4'-Fluoro-3'-methylacetophenone
Assay 97%
Molecular Formula C9H9FO
MDL Number MFCD04972879
CAS 369-32-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Odor Characteristic
Quantity 1g

Applications
4′-Fluoro-3′-methylacetophenone is used as pharmaceutical intermediate, and an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids, bases.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.