missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-methyl-5-(trifluoromethyl)phenol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1373920-61-6
Molecular Formula C8H6F4O
Molecular Weight (g/mol) 194.13
MDL Number MFCD22201069
InChI Key ZYLHFHFPFCTERO-UHFFFAOYSA-N
Synonym 4-fluoro-3-methyl-5-trifluoromethyl phenol
PubChem CID 86277676
IUPAC Name 4-fluoro-3-methyl-5-(trifluoromethyl)phenol
SMILES CC1=CC(O)=CC(=C1F)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH33276MD
View Documents Promotion Details
Thermo Scientific Chemicals
H33276MD
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1373920-61-6
194.13
ZYLHFHFPFCTERO-UHFFFAOYSA-N
86277676
CC1=CC(O)=CC(=C1F)C(F)(F)F
C8H6F4O
MFCD22201069
4-fluoro-3-methyl-5-trifluoromethyl phenol
4-fluoro-3-methyl-5-(trifluoromethyl)phenol

Specifications

1373920-61-6
MFCD22201069
UN3261
ZYLHFHFPFCTERO-UHFFFAOYSA-N
4-fluoro-3-methyl-5-(trifluoromethyl)phenol
86277676
97%
C8H6F4O
250mg
4-fluoro-3-methyl-5-trifluoromethyl phenol
CC1=CC(O)=CC(=C1F)C(F)(F)F
194.13
194.13
4-Fluoro-3-methyl-5-(trifluoromethyl)phenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'