missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride, 97%

Manufacturer:  Alfa Aesar™ H3438006

 View more versions of this product

Catalog No. AAH3438006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride
5g
97%
MFCD22201065
CVJZTMWMSKRYDY-UHFFFAOYSA-N
4-fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride
86277672
Liquid
Moisture Sensitive
1373920-95-6
C9H5ClF4O
UN3265
CC1=C(C(=CC(=C1)C(=O)Cl)C(F)(F)F)F
240.582
240.58

Chemical Identifiers

1373920-95-6
240.582
CVJZTMWMSKRYDY-UHFFFAOYSA-N
C9H5ClF4O
MFCD22201065
86277672
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents