Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester
Formula Weight 236.09
Synonym 4-Fluoro-3-(methoxycarbonyl)phenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6238503

 
alfa aesar™
H6238503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62385MD

 
alfa aesar™
H62385MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.09
Physical Form Solid
Assay 96%
Chemical Name or Material 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester
Quantity 1g
Synonym 4-Fluoro-3-(methoxycarbonyl)phenylboronic acid pinacol ester