missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5312303

 View more versions of this product

Catalog No. AAH5312303



Specifications

Specifications

4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid
1g
C8H8BFO4
4-fluoro-3-methoxycarbonyl phenylboronic acid, 4-fluoro-3-methoxycarbonylphenyl boronic acid, 4-fluoro-3-methoxycarbonyl phenyl boronic acid, 4-fluoro-3-methoxycarbonyl phenyl boronic acid, acmc-209qkz, ksc495k9r, 3-methoxycarbonyl-4-fluorophenylboronic acid, 4-fluoro-3-methoxycarbonylphenyl boronicacid, 4-fluoro-3-methoxycarbonyl-phenyl boronic acid, 4-fluoranyl-3-methoxycarbonyl-phenyl boronic acid
B(C1=CC(=C(C=C1)F)C(=O)OC)(O)O
197.956
197.96
253°C to 255°C
874219-35-9
MFCD06801688
GQBWFAIHGHHNMM-UHFFFAOYSA-N
(4-fluoro-3-methoxycarbonylphenyl)boronic acid
23005332
98%

Chemical Identifiers

874219-35-9
197.956
GQBWFAIHGHHNMM-UHFFFAOYSA-N
23005332
B(C1=CC(=C(C=C1)F)C(=O)OC)(O)O
C8H8BFO4
MFCD06801688
4-fluoro-3-methoxycarbonyl phenylboronic acid, 4-fluoro-3-methoxycarbonylphenyl boronic acid, 4-fluoro-3-methoxycarbonyl phenyl boronic acid, 4-fluoro-3-methoxycarbonyl phenyl boronic acid, acmc-209qkz, ksc495k9r, 3-methoxycarbonyl-4-fluorophenylboronic acid, 4-fluoro-3-methoxycarbonylphenyl boronicacid, 4-fluoro-3-methoxycarbonyl-phenyl boronic acid, 4-fluoranyl-3-methoxycarbonyl-phenyl boronic acid
(4-fluoro-3-methoxycarbonylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents