Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methoxybenzonitrile, 98+%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-methoxybenzonitrile
CAS 243128-37-2
Molecular Formula C8H6FNO
Formula Weight 151.14
MDL Number MFCD01569371
UN Number UN3439
Beilstein 2965042
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1989506

 
alfa aesar™
L1989506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1989514

 
alfa aesar™
L1989514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 151.14
Chemical Name or Material 4-Fluoro-3-methoxybenzonitrile
CAS 243128-37-2
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Molecular Formula C8H6FNO
MDL Number MFCD01569371
UN Number UN3439
Beilstein 2965042
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Boiling Point 96° to 98°C (1.5mmHg)
Melting Point 108° to 109°C
Quantity 5g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.