missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methoxybenzeneboronic acid pinacol ester, 96%

$53.36 - $792.35

Chemical Identifiers

CAS 425378-85-4
Molecular Formula C13H18BFO3
Molecular Weight (g/mol) 252.092
MDL Number MFCD11227055
InChI Key WAEWRICTVAIONI-UHFFFAOYSA-N
Synonym 2-4-fluoro-3-methoxy-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 22285305
IUPAC Name 2-(4-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54778MD View Documents Alfa Aesar™
H54778MD
250mg Each for $53.36
 
AAH5477803 View Documents Alfa Aesar™
H5477803
1g Each for $176.80
 
AAH5477806 View Documents Alfa Aesar™
H5477806
5g Each for $792.35
 
Specifications

Chemical Identifiers

425378-85-4
252.092
WAEWRICTVAIONI-UHFFFAOYSA-N
22285305
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)OC
C13H18BFO3
MFCD11227055
2-4-fluoro-3-methoxy-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(4-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

69°C to 71°C
425378-85-4
MFCD11227055
WAEWRICTVAIONI-UHFFFAOYSA-N
2-(4-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
22285305
96%
250mg
C13H18BFO3
2-4-fluoro-3-methoxy-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)OC
252.092
252.09
4-Fluoro-3-methoxybenzeneboronic acid pinacol ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon