missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-methoxybenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5477803

 View more versions of this product

Catalog No. AAH5477803



Specifications

Specifications

4-Fluoro-3-methoxybenzeneboronic acid pinacol ester
1g
C13H18BFO3
2-4-fluoro-3-methoxy-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)OC
252.092
252.09
69°C to 71°C
425378-85-4
MFCD11227055
WAEWRICTVAIONI-UHFFFAOYSA-N
2-(4-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
22285305
96%

Chemical Identifiers

425378-85-4
252.092
WAEWRICTVAIONI-UHFFFAOYSA-N
22285305
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)OC
C13H18BFO3
MFCD11227055
2-4-fluoro-3-methoxy-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(4-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Documents