Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-hydroxybenzoic acid, 98%

$378.20 - $378.20

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-hydroxybenzoic acid
CAS 51446-31-2
Molecular Formula C7H5FO3
Formula Weight 156.11
MDL Number MFCD00269641
Beilstein 2829723
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2534006

  SDS
alfa aesar™
B2534006
5g Each for $378.20
Description & Specifications

Specifications

Formula Weight 156.11
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 2829723
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Molecular Formula C7H5FO3
Chemical Name or Material 4-Fluoro-3-hydroxybenzoic acid
MDL Number MFCD00269641
Melting Point 215° to 220°C
Assay 98%
CAS 51446-31-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.