missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-hydroxybenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

CAS 1392234-97-7
Molecular Formula C12H16BFO3
Molecular Weight (g/mol) 238.065
InChI Key HMGMCEWHAKSNBV-UHFFFAOYSA-N
Synonym 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol
PubChem CID 68293787
IUPAC Name 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62041MD View Documents Alfa Aesar™
H62041MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6204103 View Documents Alfa Aesar™
H6204103
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1392234-97-7
238.065
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
C12H16BFO3
HMGMCEWHAKSNBV-UHFFFAOYSA-N
68293787
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)O

Specifications

250mg
C12H16BFO3
HMGMCEWHAKSNBV-UHFFFAOYSA-N
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
68293787
Solid
4-Fluoro-3-hydroxybenzeneboronic acid pinacol ester
1392234-97-7
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)O
238.065
238.06
96%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures