missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-3-formylphenylboronic acid, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445440050
Specifications
4-Fluoro-3-formylphenylboronic acid | |
5g | |
94% min. (HPLC) | |
3-formyl 4-fluorophenyl boronic acid, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid, 4-fluoro-3-formyl-phenyl boronic acid, 4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, 5-borono-2-fluorobenzaldehyde, acmc-1cswy, boronic acid, 4-fluoro-3-formylphenyl, pubchem10809 | |
B(C1=CC(=C(C=C1)F)C=O)(O)O | |
167.93 | |
167.93 |
132°C to 134°C | |
374538-01-9 | |
C7H6BFO3 | |
YABSTJQEBSKPCG-UHFFFAOYSA-N | |
(4-fluoro-3-formylphenyl)boronic acid | |
2778651 | |
95% |
Chemical Identifiers
374538-01-9 | |
167.93 | |
3-formyl 4-fluorophenyl boronic acid, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid, 4-fluoro-3-formyl-phenyl boronic acid, 4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, 5-borono-2-fluorobenzaldehyde, acmc-1cswy, boronic acid, 4-fluoro-3-formylphenyl, pubchem10809 | |
(4-fluoro-3-formylphenyl)boronic acid |
C7H6BFO3 | |
YABSTJQEBSKPCG-UHFFFAOYSA-N | |
2778651 | |
B(C1=CC(=C(C=C1)F)C=O)(O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning