Please login to your online account to display your discounted pricing

4-Fluoro-3-formylphenylboronic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-formylphenylboronic acid
Assay Percent Range 95%
Molecular Formula C7H6BFO3
Formula Weight 167.93
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC445440050

 
thermo fisher scientific
AC445440050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC445440250

 
thermo fisher scientific
AC445440250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-Fluoro-3-formylphenylboronic acid
Quantity 5g
CAS 374538-01-9
Assay Percent Range 95%
Molecular Formula C7H6BFO3
Formula Weight 167.93