missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-formylphenylboronic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445440250

 View more versions of this product

Catalog No. AC445440250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Fluoro-3-formylphenylboronic acid
25g
94% min. (HPLC)
3-formyl 4-fluorophenyl boronic acid, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid, 4-fluoro-3-formyl-phenyl boronic acid, 4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, 5-borono-2-fluorobenzaldehyde, acmc-1cswy, boronic acid, 4-fluoro-3-formylphenyl, pubchem10809
B(C1=CC(=C(C=C1)F)C=O)(O)O
167.93
167.93
132°C to 134°C
374538-01-9
C7H6BFO3
YABSTJQEBSKPCG-UHFFFAOYSA-N
(4-fluoro-3-formylphenyl)boronic acid
2778651
95%

Chemical Identifiers

374538-01-9
167.93
3-formyl 4-fluorophenyl boronic acid, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid, 4-fluoro-3-formyl-phenyl boronic acid, 4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, 5-borono-2-fluorobenzaldehyde, acmc-1cswy, boronic acid, 4-fluoro-3-formylphenyl, pubchem10809
(4-fluoro-3-formylphenyl)boronic acid
C7H6BFO3
YABSTJQEBSKPCG-UHFFFAOYSA-N
2778651
B(C1=CC(=C(C=C1)F)C=O)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents