missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-formylbenzeneboronic acid, 98+%

$97.61 - $336.18

Chemical Identifiers

CAS 374538-01-9
Molecular Formula C7H6BFO3
Molecular Weight (g/mol) 167.93
MDL Number MFCD02093074
InChI Key YABSTJQEBSKPCG-UHFFFAOYSA-N
Synonym 4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809, acmc-1cswy, 3-formyl 4-fluorophenyl boronic acid, 5-borono-2-fluorobenzaldehyde, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid
PubChem CID 2778651
IUPAC Name (4-fluoro-3-formylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)C=O)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1780803 View Documents Alfa Aesar™
L1780803
1g Each for $97.61
 
AAL1780806 View Documents Alfa Aesar™
L1780806
5g Each for $336.18
 
Specifications

Chemical Identifiers

374538-01-9
167.93
YABSTJQEBSKPCG-UHFFFAOYSA-N
2778651
B(C1=CC(=C(C=C1)F)C=O)(O)O
C7H6BFO3
MFCD02093074
4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809, acmc-1cswy, 3-formyl 4-fluorophenyl boronic acid, 5-borono-2-fluorobenzaldehyde, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid
(4-fluoro-3-formylphenyl)boronic acid

Specifications

~240°C
374538-01-9
MFCD02093074
4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809, acmc-1cswy, 3-formyl 4-fluorophenyl boronic acid, 5-borono-2-fluorobenzaldehyde, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid
B(C1=CC(=C(C=C1)F)C=O)(O)O
167.93
167.93
4-Fluoro-3-formylbenzeneboronic acid
1g
C7H6BFO3
9046762
YABSTJQEBSKPCG-UHFFFAOYSA-N
(4-fluoro-3-formylphenyl)boronic acid
2778651
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning