Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoro-3-biphenylmethylamine hydrochloride, 99%

$124.68 - $434.08

Chemical Identifiers

Chemical Name or Material 4'-Fluoro-3-biphenylmethylamine hydrochloride
CAS 1195901-44-0
Molecular Formula C13H12FN·HCl
Formula Weight 237.71
MDL Number MFCD02089423
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5029803

 
alfa aesar™
H5029803
1g Each for $434.08

AAH50298MD

 
alfa aesar™
H50298MD
250mg Each for $124.68
Description & Specifications

Specifications

Formula Weight 237.71
CAS 1195901-44-0
Chemical Name or Material 4'-Fluoro-3-biphenylmethylamine hydrochloride
Assay 99%
Molecular Formula C13H12FN·HCl
MDL Number MFCD02089423
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.