Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3,5-dimethylbenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3,5-dimethylbenzyl bromide
CAS 886501-82-2
Molecular Formula C9H10BrF
Formula Weight 217.08
MDL Number MFCD06660268
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2671806

 
alfa aesar™
H2671806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2671803

 
alfa aesar™
H2671803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 217.08
Chemical Name or Material 4-Fluoro-3,5-dimethylbenzyl bromide
Assay 97%
Molecular Formula C9H10BrF
MDL Number MFCD06660268
CAS 886501-82-2
UN Number UN3261
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 44° to 46°C
Quantity 5g

Applications
4-Fluoro-3,5-dimethylbenzyl bromide is used as laboratory chemical. It acts as an intermediate in the pharmaceutical industry.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.