Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3,5-dimethylaniline, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3,5-dimethylaniline
CAS 1840-27-3
Molecular Formula C8H10FN
Formula Weight 139.17
MDL Number MFCD09832247
UN Number UN2811
Synonym 4-Fluoro-3,5-xylidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH31663MD

  SDS
alfa aesar™
H31663MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3166303

  SDS
alfa aesar™
H3166303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.17
Melting Point 30° to 32°C
MDL Number MFCD09832247
Health Hazard 1 Danger
Assay 97%
CAS 1840-27-3
Synonym 4-Fluoro-3,5-xylidine
Health Hazard 2 GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H10FN
Chemical Name or Material 4-Fluoro-3,5-dimethylaniline
UN Number UN2811
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 250mg

Applications
4-Fluoro-3,5-dimethylaniline is used in the novel synthesis of fluoroaromatics.

Solubility
Sparingly soluble in water 0.71 g/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.