missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(2-pyridylcarbamoyl)benzeneboronic acid, 97%

$124.69 - $397.09

Chemical Identifiers

CAS 1449131-72-9
Molecular Formula C12H10BFN2O3
Molecular Weight (g/mol) 260.031
MDL Number MFCD20040156
InChI Key DJNCGHBHBJUZKA-UHFFFAOYSA-N
Synonym 4-fluoro-3-2-pyridylcarbamoyl benzeneboronic acid, 4-fluoro-3-pyridin-2-yl carbamoyl phenylboronic acid
PubChem CID 73995690
IUPAC Name [4-fluoro-3-(pyridin-2-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)F)C(=O)NC2=CC=CC=N2)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54210MD
View Documents
Alfa Aesar™
H54210MD
250mg
Each for $124.69
 
AAH5421003
View Documents
Alfa Aesar™
H5421003
1g
Each for $397.09
 
Description

Description

Specifications

Chemical Identifiers

1449131-72-9
260.031
DJNCGHBHBJUZKA-UHFFFAOYSA-N
73995690
B(C1=CC(=C(C=C1)F)C(=O)NC2=CC=CC=N2)(O)O
C12H10BFN2O3
MFCD20040156
4-fluoro-3-2-pyridylcarbamoyl benzeneboronic acid, 4-fluoro-3-pyridin-2-yl carbamoyl phenylboronic acid
[4-fluoro-3-(pyridin-2-ylcarbamoyl)phenyl]boronic acid

Specifications

250mg
C12H10BFN2O3
4-fluoro-3-2-pyridylcarbamoyl benzeneboronic acid, 4-fluoro-3-pyridin-2-yl carbamoyl phenylboronic acid
B(C1=CC(=C(C=C1)F)C(=O)NC2=CC=CC=N2)(O)O
260.031
260.03
4-Fluoro-3-(2-pyridylcarbamoyl)benzeneboronic acid
1449131-72-9
MFCD20040156
DJNCGHBHBJUZKA-UHFFFAOYSA-N
[4-fluoro-3-(pyridin-2-ylcarbamoyl)phenyl]boronic acid
73995690
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS