missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 4-Fluoro-3-(2-pyridylcarbamoyl)benzeneboronic acid, 97%
Manufacturer: Alfa Aesar™ H54210MD
Specifications
4-Fluoro-3-(2-pyridylcarbamoyl)benzeneboronic acid | |
1449131-72-9 | |
MFCD20040156 | |
DJNCGHBHBJUZKA-UHFFFAOYSA-N | |
[4-fluoro-3-(pyridin-2-ylcarbamoyl)phenyl]boronic acid | |
73995690 | |
97% |
250mg | |
C12H10BFN2O3 | |
4-fluoro-3-2-pyridylcarbamoyl benzeneboronic acid, 4-fluoro-3-pyridin-2-yl carbamoyl phenylboronic acid | |
B(C1=CC(=C(C=C1)F)C(=O)NC2=CC=CC=N2)(O)O | |
260.031 | |
260.03 |
Chemical Identifiers
1449131-72-9 | |
260.031 | |
DJNCGHBHBJUZKA-UHFFFAOYSA-N | |
73995690 | |
B(C1=CC(=C(C=C1)F)C(=O)NC2=CC=CC=N2)(O)O |
C12H10BFN2O3 | |
MFCD20040156 | |
4-fluoro-3-2-pyridylcarbamoyl benzeneboronic acid, 4-fluoro-3-pyridin-2-yl carbamoyl phenylboronic acid | |
[4-fluoro-3-(pyridin-2-ylcarbamoyl)phenyl]boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures