Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-(trifluoromethyl)cinnamic acid, 97+%

$90.05 - $352.50

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)cinnamic acid
CAS 243977-21-1
Molecular Formula C10H6F4O2
Formula Weight 234.15
MDL Number MFCD00236297
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2280603

 
alfa aesar™
B2280603
1g Each for $90.05

AAB2280606

 
alfa aesar™
B2280606
5g Each for $352.50
Description & Specifications

Specifications

Formula Weight 234.15
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)cinnamic acid
Assay ≥97%
Molecular Formula C10H6F4O2
MDL Number MFCD00236297
CAS 243977-21-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 181° to 183°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.