missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-2-(trifluoromethyl)benzoyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC398800050

 View more versions of this product

Catalog No. AC398800050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
226.56
Clear colorless to yellow
100.0
4-Fluoro-2-(trifluoromethyl)benzoyl chloride
MFCD00061158
97%
Glass bottle
Liquid
5g
1.495g/mL
96.0
189807-21-4
C8H3ClF4O
CF3C6H3(F)COCl
Authentic

Chemical Identifiers

189807-21-4
100.0
C8H3ClF4O
226.56
4-Fluoro-2-(trifluoromethyl)benzoyl chloride
96.0
97%
CF3C6H3(F)COCl
MFCD00061158
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger