Learn More
4-Fluoro-2-(trifluoromethyl)benzoyl chloride, 97%, ACROS Organics™
Chemical Identifiers
CAS | 189807-21-4 |
---|---|
Molecular Formula | C8H3ClF4O |
Molecular Weight (g/mol) | 226.56 |
MDL Number | MFCD00061158 |
InChI Key | OEYHURRIOWWRMD-UHFFFAOYSA-N |
Synonym | 2-chlorocarbonyl-5-fluorobenzotrifluoride, 2-trifluoromethyl-4-fluorobenzoyl chloride, 2-trifluoromethyl-4-fluorobenzoylchlorid, 4-fluoro-2-trifluoromethyl benzoyl chloride, 4-fluoro-2-trifluoromethyl benzoylchloride, acmc-1bwn8, alpha,alpha,alpha,4-tetrafluoro-o-toluoyl chloride, attercop-chm at107069, benzoyl chloride, 4-fluoro-2-trifluoromethyl, ksc540e2h |
PubChem CID | 605680 |
IUPAC Name | 4-fluoro-2-(trifluoromethyl)benzoyl chloride |
SMILES | C1=CC(=C(C=C1F)C(F)(F)F)C(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC398800010
|
Thermo Fisher Scientific
AC398800010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC398800050
|
Thermo Fisher Scientific
AC398800050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
189807-21-4 | |
226.56 | |
OEYHURRIOWWRMD-UHFFFAOYSA-N | |
605680 | |
C1=CC(=C(C=C1F)C(F)(F)F)C(=O)Cl |
C8H3ClF4O | |
MFCD00061158 | |
2-chlorocarbonyl-5-fluorobenzotrifluoride, 2-trifluoromethyl-4-fluorobenzoyl chloride, 2-trifluoromethyl-4-fluorobenzoylchlorid, 4-fluoro-2-trifluoromethyl benzoyl chloride, 4-fluoro-2-trifluoromethyl benzoylchloride, acmc-1bwn8, alpha,alpha,alpha,4-tetrafluoro-o-toluoyl chloride, attercop-chm at107069, benzoyl chloride, 4-fluoro-2-trifluoromethyl, ksc540e2h | |
4-fluoro-2-(trifluoromethyl)benzoyl chloride |
Specifications
1.4950g/mL | |
Authentic | |
1.495 | |
Colorless to Yellow | |
189807-21-4 | |
100.0 | |
C8H3ClF4O | |
MFCD00061158 | |
OEYHURRIOWWRMD-UHFFFAOYSA-N | |
4-fluoro-2-(trifluoromethyl)benzoyl chloride | |
605680 | |
Liquid | |
4-Fluoro-2-(trifluoromethyl)benzoyl chloride |
85°C | |
Glass bottle | |
157.0°C | |
1g | |
96.0 | |
96% min. (GC) | |
CF3C6H3(F)COCl | |
2-chlorocarbonyl-5-fluorobenzotrifluoride, 2-trifluoromethyl-4-fluorobenzoyl chloride, 2-trifluoromethyl-4-fluorobenzoylchlorid, 4-fluoro-2-trifluoromethyl benzoyl chloride, 4-fluoro-2-trifluoromethyl benzoylchloride, acmc-1bwn8, alpha,alpha,alpha,4-tetrafluoro-o-toluoyl chloride, attercop-chm at107069, benzoyl chloride, 4-fluoro-2-trifluoromethyl, ksc540e2h | |
C1=CC(=C(C=C1F)C(F)(F)F)C(=O)Cl | |
226.56 | |
226.56 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger