Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-(trifluoromethyl)benzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzoyl chloride
CAS 189807-21-4
Molecular Formula C8H3ClF4O
Formula Weight 226.56
MDL Number MFCD00061158
UN Number UN3265
Synonym alpha,alpha,alpha,4-Tetrafluoro-o-toluoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2006403

  SDS
alfa aesar™
B2006403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2006406

  SDS
alfa aesar™
B2006406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.56
Flash Point 85°C (185°F)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzoyl chloride
UN Number UN3265
CAS 189807-21-4
Boiling Point 157°C
Assay 97%
Synonym alpha,alpha,alpha,4-Tetrafluoro-o-toluoyl chloride
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
MDL Number MFCD00061158
Molecular Formula C8H3ClF4O

Applications
4-Fluoro-2-(trifluoromethyl)benzoyl chloride is used in medicine.

Solubility
Not miscible in water.

Notes
Store at 4°C. Store away from moisture and oxidizing agents under dry inert gas. Incompatible with active metals, bases and amines. Moisture Sensitive.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.