Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoro-2'-(trifluoromethyl)acetophenone, 97%

$78.04 - $78.04

Chemical Identifiers

Chemical Name or Material 4'-Fluoro-2'-(trifluoromethyl)acetophenone
CAS 208173-21-1
Molecular Formula C9H6F4O
Formula Weight 206.14
MDL Number MFCD00061251
Beilstein 7637196
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2055803

 
alfa aesar™
B2055803
1g Each for $78.04
Description & Specifications

Specifications

Formula Weight 206.14
Chemical Name or Material 4'-Fluoro-2'-(trifluoromethyl)acetophenone
Assay 97%
Molecular Formula C9H6F4O
MDL Number MFCD00061251
CAS 208173-21-1
Beilstein 7637196
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.