missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-(trifluoromethoxy)phenylacetonitrile, 97%

Manufacturer:  Alfa Aesar™ H34369MD

 View more versions of this product

Catalog No. AAH34369MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Fluoro-2-(trifluoromethoxy)phenylacetonitrile
1323966-14-8
MFCD19687176
4-fluoro-2-trifluoromethoxy phenylacetonitrile, 2-4-fluoro-2-trifluoromethoxy phenyl acetonitrile, 2-4-fluoro-2-trifluoromethoxy phenyl acetonitrile
C1=CC(=C(C=C1F)OC(F)(F)F)CC#N
219.139
219.14
97%
250mg
C9H5F4NO
UN3276
BMLRJRAUOHQFRW-UHFFFAOYSA-N
2-[4-fluoro-2-(trifluoromethoxy)phenyl]acetonitrile
66523592
Liquid

Chemical Identifiers

1323966-14-8
219.139
BMLRJRAUOHQFRW-UHFFFAOYSA-N
66523592
C1=CC(=C(C=C1F)OC(F)(F)F)CC#N
C9H5F4NO
MFCD19687176
4-fluoro-2-trifluoromethoxy phenylacetonitrile, 2-4-fluoro-2-trifluoromethoxy phenyl acetonitrile, 2-4-fluoro-2-trifluoromethoxy phenyl acetonitrile
2-[4-fluoro-2-(trifluoromethoxy)phenyl]acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

SDS Documents