Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-trifluoromethoxy-DL-phenylalanine, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-trifluoromethoxy-DL-phenylalanine
Formula Weight 267.18
MDL Number MFCD19687192
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3405103

  SDS
alfa aesar™
H3405103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH34051MD

  SDS
alfa aesar™
H34051MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.18
Chemical Name or Material 4-Fluoro-2-trifluoromethoxy-DL-phenylalanine
Assay 97%
Physical Form Solid
Quantity 1g
MDL Number MFCD19687192