missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-(trifluoromethoxy)benzyl alcohol, 97%

Manufacturer:  Alfa Aesar™ H3436203

 View more versions of this product

Catalog No. AAH3436203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Fluoro-2-(trifluoromethoxy)benzyl alcohol
1242252-68-1
C8H6F4O2
QTZPHLXETXULFE-UHFFFAOYSA-N
[4-fluoro-2-(trifluoromethoxy)phenyl]methanol
66523583
Solid
1g
97%
MFCD19687174
C1=CC(=C(C=C1F)OC(F)(F)F)CO
210.128
210.13

Chemical Identifiers

1242252-68-1
210.128
QTZPHLXETXULFE-UHFFFAOYSA-N
C8H6F4O2
MFCD19687174
66523583
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents