missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-2-nitrotoluene, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 119520250
Specifications
4-Fluoro-2-nitrotoluene | |
Authentic | |
1.5218 to 1.5238 | |
Amber to Yellow | |
25g | |
446-10-6 | |
100.0 | |
C7H6FNO2 | |
MFCD00007200 | |
2-nitro-4-fluorotoluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoro-2-nitro toluene, 4-fluoro-2-nitrobenzene, 4-fluoro-2-nitrotoluene, 4-fluoronitrotoluene, 5-fluoro-2-methylnitrobenzene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, zlchem 482 | |
CC1=C(C=C(C=C1)F)[N+](=O)[O-] | |
155.13 | |
155.13 | |
98% |
98°C | |
Glass bottle | |
138°C to 139°C (83.0mmHg) | |
27°C | |
98% | |
97.5 | |
97.5% min. (GC) | |
CH3C6H3(NO2)F | |
05, I, 161 | |
SKWTUNAAJNDEIK-UHFFFAOYSA-N | |
4-fluoro-1-methyl-2-nitrobenzene | |
67965 | |
Liquid |
Chemical Identifiers
446-10-6 | |
155.13 | |
SKWTUNAAJNDEIK-UHFFFAOYSA-N | |
67965 | |
CC1=C(C=C(C=C1)F)[N+](=O)[O-] |
C7H6FNO2 | |
MFCD00007200 | |
2-nitro-4-fluorotoluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoro-2-nitro toluene, 4-fluoro-2-nitrobenzene, 4-fluoro-2-nitrotoluene, 4-fluoronitrotoluene, 5-fluoro-2-methylnitrobenzene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, zlchem 482 | |
4-fluoro-1-methyl-2-nitrobenzene |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 207-161-8