missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-2-nitrotoluene, 98%, ACROS Organics™

Chemical Identifiers

CAS 446-10-6
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00007200
InChI Key SKWTUNAAJNDEIK-UHFFFAOYSA-N
Synonym 2-nitro-4-fluorotoluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoro-2-nitro toluene, 4-fluoro-2-nitrobenzene, 4-fluoro-2-nitrotoluene, 4-fluoronitrotoluene, 5-fluoro-2-methylnitrobenzene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, zlchem 482
PubChem CID 67965
IUPAC Name 4-fluoro-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)F)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119520100
SDS View Documents
Thermo Fisher Scientific
119520100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC119520250
SDS View Documents
Thermo Fisher Scientific
119520250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

446-10-6
155.13
SKWTUNAAJNDEIK-UHFFFAOYSA-N
67965
CC1=C(C=C(C=C1)F)[N+](=O)[O-]
C7H6FNO2
MFCD00007200
2-nitro-4-fluorotoluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoro-2-nitro toluene, 4-fluoro-2-nitrobenzene, 4-fluoro-2-nitrotoluene, 4-fluoronitrotoluene, 5-fluoro-2-methylnitrobenzene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, zlchem 482
4-fluoro-1-methyl-2-nitrobenzene

Specifications

98°C
Glass bottle
138°C to 139°C (83.0mmHg)
27°C
446-10-6
100.0
C7H6FNO2
MFCD00007200
2-nitro-4-fluorotoluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoro-2-nitro toluene, 4-fluoro-2-nitrobenzene, 4-fluoro-2-nitrotoluene, 4-fluoronitrotoluene, 5-fluoro-2-methylnitrobenzene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, zlchem 482
CC1=C(C=C(C=C1)F)[N+](=O)[O-]
155.13
155.13
98%
Authentic
1.5218 to 1.5238
Amber to Yellow
10g
97.5
97.5% min. (GC)
CH3C6H3(NO2)F
05, I, 161
SKWTUNAAJNDEIK-UHFFFAOYSA-N
4-fluoro-1-methyl-2-nitrobenzene
67965
Liquid
4-Fluoro-2-nitrotoluene, 98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 207-161-8

SDS