Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-nitrotoluene, 98%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-nitrotoluene
CAS 446-10-6
Molecular Formula C7H6FNO2
Formula Weight 155.13
MDL Number MFCD00007200
UN Number UN2811
Beilstein 1946051
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2467706

  SDS
alfa aesar™
B2467706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2467714

  SDS
alfa aesar™
B2467714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2467722

  SDS
alfa aesar™
B2467722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.13
Chemical Name or Material 4-Fluoro-2-nitrotoluene
Assay 98%
Molecular Formula C7H6FNO2
CAS 446-10-6
Beilstein 1946051
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 137° to 139°C (83mmHg)
Melting Point 25° to 27°C
Quantity 5g
MDL Number MFCD00007200
UN Number UN2811
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Flash Point 98°C (208°F)
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.