missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrophenol 98.0+%, TCI America™

Supplier:  TCI America F036725G

 View more versions of this product

Catalog No. F036725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Fluoro-2-nitrophenol
75°C
MFCD00042528
2-nitro-4-fluorophenol, phenol, 4-fluoro-2-nitro, phenol,2-nitro,4-fluoro, 4-fluoro-2-nitro-phenol, 4-fluoronitrophenol, pubchem1503, acmc-1ah4y, ksc493s6p, 5-fluoro-2-hydroxynitrobenzene, 4-fluoro-2-nitrophenol
C1=CC(=C(C=C1F)[N+](=O)[O-])O
157.1
157.10
Crystalline Powder
394-33-2
C6H4FNO3
25 g
ZHRLVDHMIJDWSS-UHFFFAOYSA-N
4-fluoro-2-nitrophenol
136236
≥98.0% (GC)

Chemical Identifiers

394-33-2
157.1
ZHRLVDHMIJDWSS-UHFFFAOYSA-N
136236
C1=CC(=C(C=C1F)[N+](=O)[O-])O
C6H4FNO3
MFCD00042528
2-nitro-4-fluorophenol, phenol, 4-fluoro-2-nitro, phenol,2-nitro,4-fluoro, 4-fluoro-2-nitro-phenol, 4-fluoronitrophenol, pubchem1503, acmc-1ah4y, ksc493s6p, 5-fluoro-2-hydroxynitrobenzene, 4-fluoro-2-nitrophenol
4-fluoro-2-nitrophenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions