missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-2-nitrobenzonitrile, 97%, Maybridge

$375.11 - $375.11

Chemical Identifiers

CAS 80517-21-1
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C7H3FN2O2
Formula Weight 166.11
MDL Number MFCD00277447
Chemical Name or Material 4-Fluoro-2-nitrobenzonitrile
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
KM06283EA Fisher Scientific Company
KM06283EA
Amber Glass Bottle 10g Each for $375.11
Specifications Description & Specifications

Chemical Identifiers

80517-21-1
97%
166.11
4-Fluoro-2-nitrobenzonitrile
97.0
C7H3FN2O2
MFCD00277447
97%

Specifications

166.11
97.0
MFCD00277447
97%
4-Fluoro-2-nitrobenzonitrile
10g
Amber Glass Bottle
C7H3FN2O2
80517-21-1
97%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning