Please login to your online account to display your discounted pricing

4-Fluoro-2-nitrobenzonitrile, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM06283EA

 View more versions of this product

Catalog No. KM06283EA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 166.11
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00277447
Chemical Name or Material 4-Fluoro-2-nitrobenzonitrile
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Molecular Formula C7H3FN2O2
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 80517-21-1

Specifications

Chemical Name or Material 4-Fluoro-2-nitrobenzonitrile
CAS 80517-21-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H3FN2O2
Formula Weight 166.11
MDL Number MFCD00277447
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.