Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-nitrobenzoic acid, 98%

$1,405.81 - $1,405.81

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-nitrobenzoic acid
CAS 394-01-4
Molecular Formula C7H4FNO4
Formula Weight 185.11
MDL Number MFCD00024300
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2282414

  SDS
alfa aesar™
B2282414
25g Each for $1,405.81
Description & Specifications

Specifications

Formula Weight 185.11
MDL Number MFCD00024300
Assay 98%
Health Hazard 1 Warning
Melting Point 147° to 150°C
Chemical Name or Material 4-Fluoro-2-nitrobenzoic acid
CAS 394-01-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Molecular Formula C7H4FNO4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.